Thermo Scientific™ Nunc™ Cell Factory™ System Accessories Available on GSA/VA Contract for Federal Government customers only.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Manufacturer: Thermo Scientific™ 170769
 View more versions of this product
Catalog No. 12-565-42
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Cell Factory Systems (all models)
    Includes
    • HDPE connector
    • Two cover caps
    • Two blue sealing caps
    • No. 4210 bacterial air vent filter
    • Tubing and clamp
    All items are sterile.
    Description Cell Factory System Start-up Kit
    Item Description Cell factory system start-up kit
    No. per Pack 1
    No. per Case 1
    Sterile Yes
    Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
    Includes:
    • HDPE connector
    • Two cover caps
    • Two blue sealing caps
    • No. 4210 bacterial air vent filter
    • Tubing and clamp
    All items are sterile.
    Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
    Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  • Description & Specifications
Related Products
Product Content Feedback