Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Nunc™ Cell Factory™ Systems
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
$535.00 - $2810.00
Specifications
Description Original Cell Factory
View More Specs
Products 6
Catalog Number Mfr. No. Culture Area Tray Layers Price Quantity & Availability  
Catalog Number Mfr. No. Culture Area Tray Layers Price Quantity & Availability  
12-565-289
Accessories View Documents
Thermo Scientific™
139446
25280cm² 40 Case of 2 for $2,810.00
N/A
12-566-90
Accessories View Documents
Thermo Scientific™
140004
2528cm² 4 Case of 10 for $1,801.00
N/A
12-565-39
Accessories View Documents
Thermo Scientific™
164327
6320cm² 10 Case of 2 for $692.00
N/A
12-565-37
Accessories View Documents
Thermo Scientific™
165250
632cm² 1 Case of 8 for $727.50
N/A
12-565-38
Accessories View Documents
Thermo Scientific™
167695
1264cm² 2 Case of 6 for $535.00
N/A
12-565-40
Accessories View Documents
Thermo Scientific™
170009
6320cm² 10 Case of 6 for $2,080.00
N/A
Description
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
  • Ideal for adherent cells
  • Boost production efficiency with more surface area in a small footprint
  • Reduce contamination risks with a closed system
Warranty: 90 days
Specifications
Specifications
Original Cell Factory
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.