Thermo Scientific™ Nunc™ Edge 96-Well Plates

Minimize plate evaporation through the uniquely engineered design of Thermo Scientific™ Nunc™ Edge 96 Well Plate

Manufacturer: Thermo Scientific™ 167311

 View more versions of this product

Catalog No. 12-565-025

  Check Availability
Add to Cart

    Specifications

    Wells 96
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    No. per Pack 160,160
    No. per Case 160
    Surface Coating Nunclon Delta
    No. of Wells 96
    Sterile Yes
    Well Design 96 F
    Color Clear
    Lid No,No

    • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
    • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment.
    • Untreated surfaces are ideal for adding coatings in your lab
    • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
    • Customized bar coding is available

    Reduced Edge Effect

    • Virtually eliminates evaporation in lengthy incubations
    • Use all 96 wells with fewer concerns of edge effects

    Low Evaporation Permits Full-Plate Results

    • Unique design minimizes plate evaporation
    • Large evaporative buffer zones eliminate well-to-well variability
    • Overall plate evaporation rate reduced to <2% after seven days of incubation
    • Perimeter moat can be filled with 1.7mL sterile H2O or media
    • Total well volume: 400µL

    Optical Properties are Critical

    • Clear plates are engineered for optical superiority
    • Low background fluorescence
    • Extreme well flatness
    • Ideal for image-based high content applications like measuring primary neuron cultures

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit