Learn More
Thermo Scientific™ Nunc™ Edge 2.0 96-Well Plates Green Product
Reduce edge effect and minimize plate evaporation using this uniquely designed moat plate simply by filling the surrounding reservoir with media or sterile water.
Manufacturer:  Thermo Scientific™ 267427
 View more versions of this product
Catalog No. 14-387-224
Add to cart
Description
Description
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Specifications
Specifications
Flat
Clear
Polystyrene
50
1
Sterile
400 μL
With Lid
8 x 12
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
0.35 cm2/well
50
Low Flange
Individually Wrapped
Non-Treated
Standard
96