Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate Available on GSA/VA Contract for Federal Government customers only.
Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.
Manufacturer: Thermo Scientific™ 237105
 View more versions of this product
Catalog No. 12-566-09
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    No. per Case 50
    No. per Pack 1
    Sterile Yes
    Surface Coating Non-Treated
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Volume (Metric) Well 400µL
    Well Design 96 F,96 F
    Color Black
    Item Description Black; Non-Treated; w/lid; 1/50
    Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.
    • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
    • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
    • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
    • Working volume range: 50 to 250µL/well
    • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback