Thermo Scientific™ Nunc™ FluoroNunc™/LumiNunc™ 96-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Use FluoroNunc/LumiNunc 96-Well Plates for quantitative and qualitative solid phase immunoassays (e.g., FLISA) and binding assays.
Manufacturer: Thermo Scientific™ 437958
 View more versions of this product
Catalog No. 12-565-290D
  Check Availability
Add to Cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
This item is not returnable. View return policy
Related Products
Product Content Feedback