Learn More
Thermo Scientific™ Nunc™ General Long Term Storage Tubes
Contain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Manufacturer:  Thermo Scientific™ 347791
 View more versions of this product
Catalog No. 12-565-302
Add to cart
Description
Description
Contiain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Starfoot vial fits into grooves of the CryoTube Rack (Cat. No. 376589), which allows one hand operation.
  • Caps and tubes packaged separately to save handling time
  • Tubes are nonsterile
  • Economical choice for less demanding applications
Specifications
Specifications
Nunc Stor-It Tubes, external thread, round bottom
Non-sterile
1200
Starfoot, round-bottom
91 mm
4.5 mL
300
12.5 mm