Learn More
Thermo Scientific™ Nunc™ General Long Term Storage Tubes
Contain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
$776.50 - $1019.00
Specifications
Description Nunc Stor-It Tubes, external thread, round bottom
Sterility Non-sterile
Style Starfoot, round-bottom
View More Specs
Products 2
Catalog Number Mfr. No. Description Volume (Metric) No. per Pack No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) No. per Pack No. per Case Price Quantity & Availability  
12-565-301
View Documents
Thermo Scientific™
347783
Nunc Stor-It Tubes, external thread, round bottom 1.8 mL 450 1800
Case of 1800 for $1,019.00
Add to cart
12-565-302
View Documents
Thermo Scientific™
347791
Nunc Stor-It Tubes, external thread, round bottom 4.5 mL 300 1200
Case of 1200 for $776.50
Add to cart
Description
Description
Contiain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Starfoot vial fits into grooves of the CryoTube Rack (Cat. No. 376589), which allows one hand operation.
  • Caps and tubes packaged separately to save handling time
  • Tubes are nonsterile
  • Economical choice for less demanding applications
Specifications
Specifications
Nunc Stor-It Tubes, external thread, round bottom
Non-sterile
Starfoot, round-bottom