Learn More
Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes.
Manufacturer:  Thermo Scientific™ 4036
 View more versions of this product
Catalog No. 08-757-10M
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes.
  • Promote larger plaques used for efficient library screening
  • Compatible with automated systems
  • Sterile; disposable
  • Deep Petri dishes allow for longer culture periods
Ordering Information:
12 per sealed plastic sleeve.
Specifications
Specifications
60mm
Sterile
14
Petri Dish
Yes
20mm
400
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.