Thermo Scientific™ Nunc™ Low Cell Binding Microplates
Inhibit cell attachment and unwanted differentiation with Thermo Scientific™ Nunc™ Low Cell Binding Microplates, suitable for a variety of cell types.
Manufacturer: Thermo Scientific™ 145397
 View more versions of this product
Catalog No. 12-567-105
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Lid Yes,Yes
    No. of Wells 96
    No. per Case 8
    No. per Pack 1,1
    Sterile Yes
    Surface Coating Low Cell Bind
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Well Design 96 F
    Closure Yes
    Color Clear
    Wells 96
    Inhibit cell attachment and unwanted differentiation of stem cells with Thermo Scientific™ Nunc™ Low Cell Binding Microplates, suitable for a variety of cell types including embryonic stem cells, neural stem cells/neurospheres and macrophages. These low cell binding microplates are irradiated and require no hydration.
    Inhibits unwanted differentiation of stem cells. Technology is based on a polymer presenting a phosphorylcholine moiety to the cells, mimicking the surface of a cell membrane. Surface is biologically inert and biocompatible, and it contains no animal-derived components.
    • Coated with a propiertary surface that inhibits cell attachment
    • Novel surface resists protein adsorption and cell adhesion
    • Clear polystyrene
    • Irradiated
    • Suitable for a variety of cell types including embroyonic stem cells, neural stem cells/neuorspheres, macrophages
    • Facilitate formation, microscopy, and transfer of embryoid bodies
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback