Learn More
Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes
Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies such as embryoid bodies.
Manufacturer:  Thermo Scientific™ 145387
 View more versions of this product
Catalog No. 12-567-104
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies with Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes.
  • Prevents attachment of adherent cells to the surface
  • Ideal for culturing embryroid bodies, neurospheres or other anchorage-independent cell clusters
  • Sterilized by gamma-irradiation and certified to be non-pyrogenic
Includes: Lid
Description & Specifications
Specifications
Yes
Yes
24
7
1mL
Multidish
1