Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes
Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies such as embryoid bodies.
Manufacturer: Thermo Scientific™ 145383
 View more versions of this product
Catalog No. 12-567-102
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Description Multidish
    Well Count 6
    No. per Pack 1
    No. per Case 7
    Volume (Metric) Working 3mL
    Sterile Yes
    Lid Yes
    Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies with Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes.
    • Prevents attachment of adherent cells to the surface
    • Ideal for culturing embryroid bodies, neurospheres or other anchorage-independent cell clusters
    • Sterilized by gamma-irradiation and certified to be non-pyrogenic
    Includes: Lid
  • Description & Specifications
Related Products
Product Content Feedback