Learn More
Invitrogen™ eBioscience™ Nunc MaxiSorp™ flat-bottom 96 well plate Non-distribution product as customer accommodation.
High protein-binding capacity polystyrene 96 well ELISA plates that are recommended for use with eBioscience Ready-SET-Go! ELISA reagent sets, as well as other matched antibody pairs for ELISA.
Manufacturer:  Invitrogen™ 44240421
Catalog No. 50-112-3685
Add to cart
Description
Description
Low-protein binding capacity plates yield low signal or inconsistent data. Use of high protein-binding capacity polystyrene 96 well ELISA plates is essential for optimal assay performance.
Specifications
Specifications
Flat
Adsorption
Standard
96
ELISA
MaxiSorp
Hydrophilic
8 x 12