Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates
Obtain consistent results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, optimized for robotics, automated readers and liquid handling systems.
Manufacturer: Thermo Scientific™ 174907
 View more versions of this product
Catalog No. 12-566-333
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Lid Yes
    Sterile Yes
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Volume (Metric) Well 400µL
    Closure Lid
    Description F 96 well plate, HydroCell, clear, with lid, Sterile (Pack of 1, Case of 8)
    No. per Case 8
    No. per Pack 8
    Surface Coating HydroCell
    Well Design 96 F,96 F
    Color Clear
    Item Description F 96 well plate, HydroCell, clear, with lid, Sterile (Pack of 1, Case of 8)
    Obtain consistent and reliable results with Thermo Scientific™ Nunc™ MicroWell™ 96-Well Microplates, which are optimized for robotics, automated readers and liquid handling systems. These microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements.
    • Clear polystyrene has excellent optical properties making it ideal for optical measurements
    • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
    • Untreated surfaces are ideal for adding coatings in your lab
    • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
    • Plates are optimized for robotics, plate readers and liquid handlers
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback