Thermo Scientific™ Nunc™ Multidishes with HydroCell™ Surface
Inhibit the attachment of adherent cells and enable the formation of cell clusters with Thermo Scientific™ Nunc™ Multidishes with HydroCell™ Surface.
Manufacturer: Thermo Scientific™ 174909
 View more versions of this product
Catalog No. 12-566-335
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
For research and single-use only.
    Specifications
    Lid Yes,Yes
    Dish Type Multidish | Cell Culture
    Culture Area 1.9cm²
    No. of Wells 24
    No. per Case 6
    No. per Pack 1,1
    Sterile Yes
    Surface Coating HydroCell
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Well Design 24 F
    Closure Yes
    Color Clear
    Wells 24
    Inhibit the attachment of adherent cells and enable the formation of cell clusters with Thermo Scientific™ Nunc™ Multidishes with Thermo Scientific™ HydroCell™ Surface, featuring super-low cell binding capacity with premium quality.
    A covalently immobilized super hydrophilic polymer forms a thin, even layer on cultureware with HydroCell Surface, resulting in minimal cell adhesion and protein adsorption. Depending on cell type, cells can be grown as a suspension of single cells or as a suspension of cell clusters.
    • Minimal cell adhesion—high yield of cells and eliminate unwanted adhesion-induced differentiation
    • Minimal protein adsorption—high yield of cell-secreted proteins
    • Ideal for culturing embryroid bodies, neurospheres, or other anchorage-independent cell clusters
    • Sterile and non-pyrogenic
    • Customized bar coding is available
    Ordering Information:
    Individually wrapped with lid.
  • Description & Specifications
Related Products
Product Content Feedback