Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ Nunc™ Multidishes with HydroCell™ Surface
Inhibit the attachment of adherent cells and enable the formation of cell clusters with Thermo Scientific™ Nunc™ Multidishes with HydroCell™ Surface.
Manufacturer:  Thermo Scientific™ 174919
 View more versions of this product
Catalog No. 12-566-370
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
For research and single-use only.
Specifications
Description
Inhibit the attachment of adherent cells and enable the formation of cell clusters with Thermo Scientific™ Nunc™ Multidishes with Thermo Scientific™ HydroCell™ Surface, featuring super-low cell binding capacity with premium quality.
A covalently immobilized super hydrophilic polymer forms a thin, even layer on cultureware with HydroCell Surface, resulting in minimal cell adhesion and protein adsorption. Depending on cell type, cells can be grown as a suspension of single cells or as a suspension of cell clusters.
  • Minimal cell adhesion—high yield of cells and eliminate unwanted adhesion-induced differentiation
  • Minimal protein adsorption—high yield of cell-secreted proteins
  • Ideal for culturing embryroid bodies, neurospheres, or other anchorage-independent cell clusters
  • Sterile and non-pyrogenic
  • Customized bar coding is available
Ordering Information:
Individually wrapped with lid.
Description & Specifications
Specifications
Yes
Yes
9.6cm²
6
Yes
6 F
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Clear
6
1
HydroCell
Multidish | Cell Culture