Learn More
Thermo Scientific™ Nunc™ Serological Pipettes
Select size and packaging options to advance your research with complete confidence when using Thermo Scientific? Nunc? Serological Pipettes.
Manufacturer:  Thermo Scientific™ 170372
 View more versions of this product
Catalog No. 12-567-613
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Select size and packaging options to advance your research with complete confidence when using Thermo Scientific™ Nunc™ Serological Pipettes. These accurate, disposable plastic pipettes are plugged and sterilized.
  • Sterility assurance level (SAL) of 10-6
  • 100% virgin polystyrene for maximum clarity
  • Certified non-pyrogenic
  • Bold clear graduations with easy-to-read scale markings
  • Non-dripping tip for pipette precision
  • Color-coded packaging for ease in sorting and selecting the correct size
  • Packaging options that accommodate every lab
  • Individually wrapped; paper/plastic or plastic film; peel or pop through for easy opening
  • Bulk packed for frequent use; reduces packaging waste
  • Multiple sizes for use in a wide range of applications
Ordering Information:
Supplied sterile, individually packaged in easy-open paper/plastic wrapper.
Description & Specifications
Specifications
2mL
Bulk packed, plugged
Serological Pipette, 2mL, Bulk packed, plugged, 50/bag, 500/case
500
Yes
500 per case
50
Sterile