Learn More
Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps
Allow constant airflow and minimize chance of contamination by using individually packed Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Manufacturer:  Thermo Scientific™ 156753
 View more versions of this product
Catalog No. 12-566-88
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Allow constant airflow and minimize chance of contamination with Thermo Scientific™ Nunc™ Sterile Flask Replacement Filter Caps.
Nunc sterile filter caps are made with HDPE and Thermo Scientific™ GORE™ dPTFE membrane.
  • Filter membrane provides effective barrier against microorganisms
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Individually wrapped to ensure sterility
Specifications
Specifications
Filter
White
Nunc EasYFlasks 75cm2
1
Sterile
HDPE
100
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.