Learn More
Thermo Scientific™ Nunc™ Strips and Frames
Maximize heat transmission and sample recovery using Nunc Strips and Frames.
Manufacturer:  Thermo Scientific™ 232042/PRC
 View more versions of this product
Catalog No. 12-565-218N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Maximize heat transmission and sample recovery using Thermo Scientific™ Nunc Strips and Frames. Thermo Scientific Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Tape and well design reduce evaporation
  • Centrifugation eliminated when mixing reagents
  • Easy well identification
  • Minimum binding of hydrophilic molecules
  • Standard 96 well format
  • Modules are temperature resistant from -70° to +121°C
  • Space saving at the bench and in the freezer
  • Plastic waste is reduced
Description & Specifications
Specifications
Sequencing Module