Learn More
Thermo Scientific™ Nunc™ Polystyrene Roller Bottles Available on GSA/VA Contract for Federal Government customers only.
Reliably grow cells with high reproducibility by using Thermo Scientific™ Nunc™ Polystyrene Roller Bottles.
Manufacturer:  Thermo Scientific™ 184902
 View more versions of this product
Catalog No. 12-565-680
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Grow cells with high reproducibility by using highly-reliable Thermo Scientific™ Nunc™ Polystyrene Roller Bottles. Developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics.
  • One-piece seamless design: Reduces possibility of leaking through a seam
  • Shallow and deep indents: Unique ergonomic design makes the roller bottle excellent for both manual and automated handling
  • Proprietary manufacturing process: Greater impact resistance
  • Vertical expanded surfaces version: Vertical pleat orientation facilitates emptying and reduces retention of product
Recommended for:
Cell Culture Expansion, Vaccine Production, Antibody Production
Description & Specifications
Specifications
Smooth surface, deep indent
Clear
20
2/Pack, 20/Case; Double-bagged
Yes
EZ twist non-vented cap
HDPE
Polystyrene
2
850cm²
Narrow