Learn More
Thermo Scientific™ Nunc™ White 384-Well Optical Bottom Plates Available on GSA/VA Contract for Federal Government customers only.
Perform luminescence measurements and other microscopic applications with these white microplates, featuring maximum reflection and minimal autoluminescence.
Manufacturer:  Thermo Scientific™ 242763
 View more versions of this product
Catalog No. 12-565-355
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Perform luminescence measurements and other microscopic applications with Thermo Scientific™ Nunc™ White 384-Well Optical Bottom Plates. These microplates have maximum reflection and minimal autoluminescence.
Rounded-square wells provide maximum access for dispensers and readers.
  • Poly- D-Lysine and Collagen coating is ideal for cells with low adherence or growth—the uniform coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment and are ideal for adding coating in your own lab
  • Polystyrene upper structure with either a #1.5 cover glass or polystyrene base
  • Working volume: 10 to 120µL
  • Customized bar coding is available
Description & Specifications
Specifications
No
No
Polystyrene
10
Non-Treated
120µL
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
White
30
No
Optical Bottom Plate
384 F