Thermo Scientific™ Nunclon™ Sphera™ Microplates

Make your most valuable work your best work. These microplates feature a superior cell culture surface with performance that elevates stem cell and cell culture research.

Manufacturer: Thermo Scientific™ 174929

 View more versions of this product

Catalog No. 12-566-432

  Check Availability
Add to Cart

Disclaimers:


Shelf Life

  • 4 years


    Specifications

    Format 96U-Well Plate,96U-Well Plate
    No. per Case 50
    No. per Pack 5,5
    Sterile SAL 10-6, ethylene oxide gas sterilization
    Surface Coating Nunclon Sphera
    Volume (Metric) 300μL,300μL
    Certifications/Compliance Free of any animal products; nonpyrogenic, endotoxin tested (0.5 EU/mL); USP biological reactivity class VI
    Well Design Round Bottom
    Surface Culture Nunclon Sphera
    Well Shape Round Bottom

    Make your most valuable work your best work. Thermo Scientific™ Nunclon™ Sphera™ Microplates feature a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research. Offered in round and flat-bottom 96-well formats.

    Significant qualities of the Nunclon Sphera culture surface:

    Optimizing spheroid formation
    • Allows cells to grow in suspension with virtually no cell attachment
    • Supports many different cell types and their ability to generate spheroids
    • Enables consistent, repeatable cell growth
    Growing 3D cancer cultures
    • Minimizes variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumor growth, providing a better model system for studying tumor cell progression and efficacy of anticancer agents in vitro
    Promoting formation of embryoid bodies (EB)
    • Demonstrates superior quality for embryoid body formation of pluripotent stem cells with minimal spontaneous differentiation
    • Resultant cells able to differentiate into all three germ layers

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit