Learn More
Thermo Scientific™ Nuova™ Hotplates
Save space with the compact design of Thermo Scientific™ Nuova™ Hotplates. These hotplates provide temperature uniformity and have a corrosion-resistant top.
Manufacturer:  Thermo Scientific™ HP18320Q
 View more versions of this product
Catalog No. 11-496-51Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Save space with the compact design of Thermo Scientific™ Nuova™ Hotplates. These hotplates provide excellent temperature uniformity and have a corrosion-resistant top.
  • Durable die-cast aluminum case
  • Excellent corrosion resistance with porcelain-coated stainless-steel top
  • Accommodates up to 20 lb. (9.1kg) loads
  • Integral ring stand holder accommodates a 0.5 in. (1.3cm) diameter support rod
  • Demand-type thermostatic temperature control provides excellent temperature stability: ±5.0°C (9°F) at 371°C (700°F)
  • Embedded heating elements transfer heat evenly and uniformly
  • Topside drip edge protects internal components in case of spillage
Description & Specifications
Specifications
38° to 371°C
7.0 x 7.0 in.
4.5 in.
8.6 in.
11.8 in.
840w
220-240V; 840w; 3.5A
8.6 x 11.8 x 4.5 in. (21.8 x 29.9 x 11.73cm)
220/240V
17.7 x 17.7cm
11.73cm
21.8cm
29.9cm
3.5A
UL, CUL (HP18325Q and HP18320Q)
Hotplates
Accessories