Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nup133 Rat anti-Mouse, Clone: 9C2H8, MilliporeSigma™

Rat Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen Nup133
Clone 9C2H8
Concentration 0.5 mg/mL
Applications Western Blot, Immunofluorescence
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE1926100
View Documents
MilliporeSigma
MABE1926100UL
100 μL
Each for $373.00
Add to cart
 
MABE192625
View Documents
MilliporeSigma
MABE192625UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

Nup133 Monoclonal antibody specifically detects Nup133 in Mouse samples. It is validated for Western Blotting, Immunofluorescence.
Specifications

Specifications

Nup133
0.5 mg/mL
Unconjugated
Rat
Mouse
His-tagged recombinant fragment corresponding to 447 amino acids from the N-terminal half of murine Nuclear pore complex protein Nup133.
Monoclonal
Protein G purified
9C2H8
Western Blot, Immunofluorescence
Purified
RUO
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit