Learn More
Aviva Systems Biology Corp NUP43 Antibody - middle region (ARP55922_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55922_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUP43. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NUP43. Alternate names: FLJ13287, bA350J20.1, p42ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61664