Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP62 Mouse anti-Human, FITC, Clone: 252, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243589F

Catalog No. NBP243589F


Add to cart

Description

Description

NUP62 Monoclonal antibody specifically detects NUP62 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NUP62
252
Immunocytochemistry/Immunofluorescence
Purified
NUP62
Recombinant protein encompassing a sequence within the center region of human Nucleoporin p62. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62
Mouse
IgG2b
0.1 ml
Primary
23636
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit