Learn More
NUPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 199071AP
Catalog No. 50-173-2594
Add to cart
Specifications
Specifications
NUPL1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70581, Q8R332, Q9BVL2
KIAA0410, nucleoporin like 1, Nucleoporin like protein 1, Nucleoporin p58/p45, NUPL1, PRO2463
Rabbit
IgG
150 μL
-20°C
Primary
245922, 71844, 9818
Western Blot
Unconjugated
NUP58
Liquid
NUP58, Nupl1
NUPL1 Fusion Protein Ag13901
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat