Learn More
Cayman Chemical nuropeptIde YporcInbovI 10mg
SDP
Supplier:  Cayman Chemical 3671510 MG
Encompass_Preferred
A neuropeptide and an NPY receptor agonist; selectively inhibits forskolin-induced cAMP accumulation in L-M(TK-) cells expressing rat Y1 or Y2 and HEK293 cells expressing rat Y5 over L-M(TK-) cells expressing rat Y4 receptors (EC50s = 0.15, 2.7, 0.66, and >1,000 nM, respectively); increases perfusion pressure in isolated guinea pig heart (EC30 = 1.3 nM); intracerebroventricular administration increases food intake in rats at 300 pmol/animal
Catalog No. 50-290-9621