Learn More
MilliporeSigma™ Nus Tag Monoclonal Antibody,
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ 711273
Catalog No. 71-127-3MI
Add to cart
Description
Description
The Nus Tag™ Monoclonal Antibody is a mouse monoclonal antibody (IgG1) with high affinity for the 54.8kDa NusA protein from E. coli. This purified antibody specifically detects fusion proteins containing the Nus Tag sequence expressed with the pET-43.1 and pET-44 vector series. Expression of polypeptides fused with the 495-aa NusA (Nus Tag) can markedly enhance the solubility of recombinant proteins expressed in E. coli (Harrison 2000, Davis 1998).
Specifications
Specifications
Nus Tag
Unconjugated
Liquid
IgG1
RUO
Primary
495aa Nus Tag Protein, precise epitope not determined
Western Blot
Stabilized solution (1mg/mL) in 50% glycerol
Mouse
50μg
-20°C
Monoclonal
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.