Learn More
NUS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 159861AP
Catalog No. 50-173-0622
Add to cart
Description
Description
With DHDDS, forms the dehydrodolichyl diphosphate synthase (DDS) complex, an essential component of the dolichol monophosphate (Dol-P) biosynthetic machinery. Adds multiple copies of isopentenyl pyrophosphate (IPP) to farnesyl pyrophosphate (FPP) to produce dehydrodolichyl diphosphate (Dedol-PP), a precursor of dolichol which is utilized as a sugar carrier in protein glycosylation in the endoplasmic reticulum (ER). Regulates the glycosylation and stability of nascent NPC2, thereby promoting trafficking of LDL-derived cholesterol. Acts as a specific receptor for the N-terminus of Nogo-B, a neural and cardiovascular regulator.
Specifications
Specifications
NUS1
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96E22, Q99LJ8
C6orf68, NgBR, Nogo B receptor, NUS1
Rabbit
IgG
150 μL
-20°C
Primary
116150, 294400, 52014
Western Blot
Unconjugated
NUS1
Liquid
Nus1
NUS1 Fusion Protein Ag8789
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat