Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Nutlin-3 Inhibitor

Low molecular weight, organic compound that may help regulate biological process.

Specifications

CAS 548472-68-0
Formula Weight 581.49g/mol
For Use With (Application) Inhibits apoptosis
Molecular Formula C30H30Cl2N4O4
Solubility Soluble to 100mM in DMSO, 100mM in ethanol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB398410MG View Documents Novus Biologicals™
NB398410MG
50mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
NB398450MG View Documents Novus Biologicals™
NB398450MG
50mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Biological Activity: MDM2 antagonist; inhibits the MDM2-p53 interaction (IC50 = 0.09 µM) and activates p53. Antiproliferative agent; induces apoptosis in cancer cells.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Specifications

548472-68-0
Inhibits apoptosis
Soluble to 100mM in DMSO, 100mM in ethanol
Nutlin-3
581.49g/mol
C30H30Cl2N4O4
Store at 4°C
White

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit