Learn More
Thermo Scientific™ Nutrient Broth Available on GSA/VA Contract for Federal Government customers only.
Reduce media preparation time and cultivate fewer fastidious microorganisms using this convenient, pre-filled, ready-to-use general purpose medium.
$215.78 - $927.15
Specifications
Color Clear liquid, light amber
Format Tube
Fill Volume 5 mL
Manufacturing Origin United States
Packaging Glass tube with black screw cap
View More Specs
Products 2
Catalog Number Mfr. No. Description Format Quantity Manufacturing Origin Price Quantity & Availability  
Catalog Number Mfr. No. Description Format Quantity Manufacturing Origin Price Quantity & Availability  
R061580
Accessories View Documents
Thermo Scientific™
R061580
Nutrient Broth Tube 100/Pk. United States
Case of 100 for $927.15
Add to cart
R061582 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R061582
Nutrient Broth Tube 20/Pk. United States
Pack of 20 for $215.78
Add to cart
Description
Description
Reduce media preparation time and cultivate fewer fastidious microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Nutrient Broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Selenite Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clear liquid, light amber
5 mL
Glass tube with black screw cap
Tube
United States
2-8°C