Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Tocris Bioscience™ NVP CXCR2 20 Available on GSA/VA Contract for Federal Government customers only.

Potent and selective CXCR2 antagonist

$159.00 - $669.00

Specifications

Chemical Name or Material 4-Cyclopropyl-2-[[(2,3-difluorophenyl)methyl]thio]-1,6-dihydro-6-oxo5-pyrimidinecarbonitrile
Molecular Formula C15H11F2N3OS
CAS 1029521-30-9
Product Type Target: Chemokine Receptor Antagonists
Formula Weight 319.33
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
56-601-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5660/10
10mg Each for $159.00
N/A
56-605-0 View Documents Tocris Bioscience™
5660/50
50mg Each for $669.00
N/A
Description

Description

Potent and selective CXCR2 antagonist (IC50 = 40 nM). Exhibits selectivity for CXCR2 over a panel of 49 other GPCRs. Orally bioavailable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

4-Cyclopropyl-2-[[(2,3-difluorophenyl)methyl]thio]-1,6-dihydro-6-oxo5-pyrimidinecarbonitrile
1029521-30-9
319.33
C15H11F2N3OS
Target: Chemokine Receptor Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit