Learn More
NWD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 250251AP
Catalog No. 50-173-4678
Add to cart
Description
Description
NWD1 also named as NWD1 Ens or NACHT and WD repeat domain containing 1 is a 1564 amino acid protein, which contains 1 NACHT domain and 13 WD repeat. The molecular weight of NWD1 was detected by western blot. The result shows that the molecular weight of NWD1 is 77 kDa., but NWD1 is a 174 kDa protein according to UniProt,. The function of NWD1 is still non-known, meanwhile the molecular weight of NWD1 is still remained to be confirmed.
Specifications
Specifications
NWD1
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A6H603, Q149M9
NWD1, NWD1 Ens
Rabbit
IgG
150 μL
-20°C
Primary
284434, 319555
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NWD1
Liquid
Nwd1
NWD1 Fusion Protein Ag21754
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse