Learn More
Aviva Systems Biology Corp NXF1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP40638_P050
This is a rabbit polyclonal antibody against NXF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NXF1. Alternate names: DKFZp667O0311, MEX67, TAPApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53218