Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551681
Description
Immunogen sequence: TAPYSVKNKL KPGQMEMLKL TMNKRYNVSQ QALDLQNLRF DPDLMGRDID IILNRRNCMA ATLKITERNF PELLSLNLCN NKLYQLDGLS DITEKAPKVK TLNLSKNKLE SAWELGKVKG LKLEELWLEG NPLCSTFSDQ S Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 68%.
This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene.
Specifications
NXF2 | |
Polyclonal | |
Unconjugated | |
NXF2 | |
4930455N06Rik; cancer/testis antigen 39; CT39; Nuclear RNA export factor 2; nuclear RNA export factor 5; NXF2; NXF2B; Nxf-b; RGD1559530; TAPL2; TAPL-2; TAP-like protein 2; t-complex protein 11 homolog; TCP11X2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZY0 | |
NXF2 | |
Recombinant protein corresponding to Human NXF2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction