Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548382

Catalog No. PA548382

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with xenopus based on sequence homology.

Functions as a redox-dependent negative regulator of the Wnt signaling pathway, possibly by preventing ubiquitination of DVL3 by the BCR(KLHL12) complex. May also function as a transcriptional regulator act as a regulator of protein phosphatase 2A (PP2A).
Specifications

Specifications

NXN
Polyclonal
PBS with 0.09% sodium azide
Q6DKJ4, P97346
Nucleoredoxin, NXN, NRX
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18230, 64359
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NXN
Liquid
NXN
KLH conjugated synthetic peptide between 221-249 amino acids from the Central region of human NXN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit