Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXNL2 Mouse anti-Human, Non-human primate, Clone: OTI2B11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525121

Catalog No. PIMA525121


Add to cart

Description

Description

May be involved in the maintece of both the function and the viability of sensory neurons, including photoreceptors and olfactory neurons.
Specifications

Specifications

NXNL2
Monoclonal
0.63 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q5VZ03
C9orf121, RDCVF2, nucleoredoxin-like protein 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
158046, 698533
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
OTI2B11
Unconjugated
NXNL2
Liquid
NXNL2
Full length human recombit protein of NXNL2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit