Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ NXY 059

Free radical trapping agent; neuroprotectant

Manufacturer:  Tocris™ 3645/10

 View more versions of this product

Catalog No. 36-451-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Free radical trapping agent. Reduces infarct size and preserves brain function in several animal models of acute ischemic stroke. Neuroprotectant.
  • Chemical Name: 4-[[(1,1-Dimethylethyl)oxoimino]methyl]-1,3-benzenedisulfonic acid disodium salt
  • Formula: C11H13NNa2O7S2
  • CAS Number: 168021-79-2
  • Solubility: Soluble to 100mM in water and to 100mM in DMSO
  • Purity: >98
Description & Specifications

Specifications

NXY 059
>98
10mg
NXY 059
381.33
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit