Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ NY-BR-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ NY-BR-1, Mouse Monoclonal Antibody.

$406.50 - $1630.00

Specifications

Antigen NY-BR-1
Clone NY-BR-1 No.2
Applications Immunohistochemistry
Conjugate Unconjugated
Host Species Murine
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1932P0
View Documents
Thermo Scientific™
MS1932P0
100μL
Each for $406.50
Add to cart
 
MS1932P
View Documents
Thermo Scientific™
MS1932P
1mL
Each for $1,630.00
Add to cart
 
MS1932R7
View Documents
Thermo Scientific™
MS1932R7
7mL
Each for $499.00
Add to cart
 
MS1932RQ
View Documents
Thermo Scientific™
MS1932RQ
12mL
Each for $682.50
Add to cart
 
MS1932R1
View Documents
Thermo Scientific™
MS1932R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ NY-BR-1, Mouse Monoclonal Antibody.

NY-BR-1 is a novel differentiation marker predominantly expressed in normal breast and breast cancer. It has been shown by Immunohistochemistry (IHC) that NY-BR-1 was present solely in ductal epithelium of normal breast tissue. Invasive carcinoma of the breast and carcinoma in situ were positive for NY-BR-1, whereas most other tumors and normal tissues are negative, with the exceptions of one-third of sweat gland carcinomas and around two percent of prostate carcinoma. NY-BR-1 is reported to express more frequently in estrogen-positive and lymph node-negative primary carcinomas, and more commonly in grade 1 than in grade 2 or grade 3. In a study by Varga et al (2006) forty-nine percent of lymph node metastasis were shown to be positive.

Host Species: Mouse

Clone: NY-BR-1No. 2

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: a 77 aa peptide from human NYBR-1 protein

Molecular Weight: 150-160kDa

Positive Control: Breast carcinoma

Cellular Localization: Predominantly cytoplasmic and occasional nuclear

Recommended for:

Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

NY-BR-1
Immunohistochemistry
Murine
a 77 aa peptide from human NYBR-1 protein
NY-BR-1 No.2
Unconjugated
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit