Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Uniflo™ 25 Syringe Filters
Offers good chemical resistance to most common HPLC solvents. Whatman™ Uniflo Syringe Filter, Nylon Membrane is a disposable, hydrophilic filter unit that is a good choice for aqueous and aqueous-organic samples for use in HPLC sample filtration.
Specifications
Material Nylon membrane
Housing Material Polypropylene
Membrane Hydrophilic
Sterility Non-sterile
Disposable Yes
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Membrane Pore Size No. per Pack Price Quantity & Availability  
09-927-431
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463872
13mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-327
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463877
13mm 0.2μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-440
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463879
13mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-587
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463433
25mm 0.2μm 1000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
09-927-326
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
10463442
25mm 0.45μm 500 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Offers good chemical resistance to most common HPLC solvents. Whatman™ Uniflo Syringe Filter, Nylon Membrane is a disposable, hydrophilic filter unit that is a good choice for aqueous and aqueous-organic samples for use in HPLC sample filtration.
Whatman Uniflo Syringe Filters are disposable filter units designed to provide clean filtrate from small volumes up to 10mL (13mm filters) or 100mL (25mL filters).
  • Combines premium quality with economic efficiency
  • Available in a variety of membrane choices with a polypropylene overmold housing
  • A range of sizes and pore sizes to meet the filtration application needs
  • Individual ink-jet printing on the filter for easy identification
  • Packaged in a clean, compact recyclable plastic box to optimize benchtop space
Specifications
Specifications
Nylon membrane
Hydrophilic
Yes
Male Slip Luer
Aqueous and/or aqueous-organic samples
UNIFLO Syringe Filter
Polypropylene
Non-sterile
Female Luer Lock
HPLC sample filtration
Resealable, recyclable plastic boxes