Learn More
MilliporeSigma™ Nylon Plain Surface Net Filter
MilliporeSigma Nylon Plain Surface Net Filter, Pore Size: 180um, For Use With: For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring, White, Dimensions: 3m x 30cm, Filter Code: NY8H, Filter Surface: Plain
$143.20 - $585.50
Specifications
Color White
Filter Surface Plain
Filter Type Net Filter
Temperature Range (Metric) 100°C
For Use With (Application) For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring
View More Specs
Products 3
Catalog Number Mfr. No. Item Description Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Item Description Pore Size Price Quantity & Availability  
NY6H02500
View Documents
MilliporeSigma™
NY6H02500
25 mm dia. 160μm Pack for $143.20
Add to cart
NY6H00010
View Documents
MilliporeSigma™
NY6H00010
3 m x 30 cm 160μm Each for $585.50
Add to cart
NY2000010
View Documents
MilliporeSigma™
NY2000010
3 m x 30 cm 20μm Pack for $585.50
Add to cart
Description
Description
MilliporeSigma Nylon Plain Surface Net Filter, Pore Size: 180um, For Use With: For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring, White, Dimensions: 3m x 30cm, Filter Code: NY8H, Filter Surface: Plain
Specifications
Specifications
White
Net Filter
For use in Collection of algae & cells, particle analysis, large particulate filtration, prefiltration of solvents, paint monitoring
Net Filter
Plain
100°C
Nylon