Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O(1),O(3)-Bis(ethoxycarbonylmethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene, Alfa Aesar™

Click to view available options
Quantity:
0.1g
0.5g
149775-71-3
C54H72O8
849.162
MFCD00798577
YKBOZEHACZIDBO-UHFFFAOYSA-N
o 1 ,o 3-bis ethoxycarbonylmethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene,o 1 ,o 3-bis ethoxycarbonylmethyl-o 2 ,o 4-dimethyl-p-ter,o 1 ,o 3-bis carbethoxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene,2,2-dimethoxy-2',2'-bis 2-oxo-2-ethoxyethoxy 1,3'-methylene-1',3-methylene-1,3'-methylene-1',3-methylenetetrakis 5-tert-butylbenzene,ethyl 2-5,11,17,23-tetra-tert-butyl-27-2-ethoxy-2-oxoethoxy-26,28-dimethoxypentacyclo 19.3.1.1_,?.1?,__.1_?,_? octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15 26 ,16,18,21,23-dodecaen-25-yl oxy acetate
73995054
CCOC(=O)COC1=C2CC3=CC(=CC(=C3OC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OC)OCC(=O)OCC)C(C)(C)C
149775-71-3
C54H72O8
849.162
MFCD00798577
YKBOZEHACZIDBO-UHFFFAOYSA-N
o 1 ,o 3-bis ethoxycarbonylmethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene,o 1 ,o 3-bis ethoxycarbonylmethyl-o 2 ,o 4-dimethyl-p-ter,o 1 ,o 3-bis carbethoxymethyl-o 2 ,o 4-dimethyl-p-tert-butylcalix 4 arene,2,2-dimethoxy-2',2'-bis 2-oxo-2-ethoxyethoxy 1,3'-methylene-1',3-methylene-1,3'-methylene-1',3-methylenetetrakis 5-tert-butylbenzene,ethyl 2-5,11,17,23-tetra-tert-butyl-27-2-ethoxy-2-oxoethoxy-26,28-dimethoxypentacyclo 19.3.1.1_,?.1?,__.1_?,_? octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15 26 ,16,18,21,23-dodecaen-25-yl oxy acetate
73995054
CCOC(=O)COC1=C2CC3=CC(=CC(=C3OC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OC)OCC(=O)OCC)C(C)(C)C
Quantity 0.1g
Melting Point 162°C to 164°C
Formula Weight 849.16
Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene

TSCA : No

Recommended Storage : Ambient temperatures