Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%

Click to view available options
Quantity:
5g
25g
100g
94790-37-1
C11H16F6N5OP
379.247
MFCD00075445
UQYZFNUUOSSNKT-UHFFFAOYSA-N
hbtu,o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,hbtu-reagent,o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate,hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate,n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate,o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate
2733084
[benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
94790-37-1
C11H16F6N5OP
379.247
MFCD00075445
UQYZFNUUOSSNKT-UHFFFAOYSA-N
hbtu,o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,hbtu-reagent,o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate,hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate,n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate,o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate
2733084
[benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F
Applications
O-(1H-Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is used as a coupling reagent for solid-phase peptide synthesis. It has been shown to effectively suppress racemization.

Solubility
Soluble in acetonitrile.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Melting Point ~200°C (decomposition)
Quantity 5g
Beilstein 7328329
Formula Weight 379.25
Percent Purity 98%
Chemical Name or Material O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C