Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
57981-02-9
C7H5ClF5NO
249.57
MFCD00012953
HVMVKNXIMUCYJA-UHFFFAOYSA-N
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv
122307
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F
57981-02-9
C7H5ClF5NO
249.57
MFCD00012953
HVMVKNXIMUCYJA-UHFFFAOYSA-N
o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv
122307
O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride
[H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9

Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point 227.0°C
Quantity 1g
Assay Percent Range 97.5 to 102.5% (Argentometry)
Linear Formula C6F5CH2ONH2·HCl
Solubility Information Solubility in water: soluble.
Formula Weight 249.57
Percent Purity 98%
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 261-057-7