Learn More
Tocris Bioscience™ O-2093
$99.00 - $425.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
14-465 View Documents Tocris Bioscience™
1446/5
Each for $99.00
Add to cart
14-462-5R View Documents Tocris Bioscience™
1446/25
Each for $425.00
Add to cart
Description
Description
Inhibitor of anandamide uptake (IC50 = 17.3 µM) with little or no activity at CB1, CB2, TRPV1 and FAAH. Intravenous administration inhibits limb spasticity in mice with chronic relapsing experimental allergic encephalomyelitis (CREAE).