Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ O-2093

Inhibitor of anandamide uptake

Manufacturer:  Tocris Bioscience™ 1446/5

 View more versions of this product

Catalog No. 14-465


Add to cart

Description

Description

Inhibitor of anandamide uptake (IC50 = 17.3µM) with little or no activity at CB1, CB2, TRPV1 and FAAH. Intravenous administration inhibits limb spasticity in mice with chronic relapsing experimental allergic encephalomyelitis (CREAE).
Specifications

Specifications

N-(bis-3-chloro-4-hydroxybenzyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
C34H43Cl2NO3
5mg
439080-01-0
Observed MW: 584.62
Soluble in ethanol (supplied pre-dissolved in anhydrous ethanol, 5mg/mL)
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at -20°C

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit