Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Acros Organics

Click to view available options
Quantity:
10g
50g
330645-87-9
C11H15ClN5O·F6P
413.69
ZHHGTMQHUWDEJF-UHFFFAOYSA-N
hctu,o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v,1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium hexafluorophosphate,2-6-chloro-1h-benzotriazole-1-yl-1,1,3,3-tetramethylaminium hexafluorophosphate,o-1h-6-chlorobenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,o-6-chloro-1-hydrocibenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v,pubchem12740,acmc-20aj2u
42624899
[(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1.F[P-](F)(F)(F)(F)F
330645-87-9
C11H15ClN5O·F6P
413.69
ZHHGTMQHUWDEJF-UHFFFAOYSA-N
hctu,o-6-chlorobenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate,2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v,1-bis-dimethylamino-methylene-5-chloro-3-oxy-1h-benzotriazol-1-ium hexafluorophosphate,2-6-chloro-1h-benzotriazole-1-yl-1,1,3,3-tetramethylaminium hexafluorophosphate,o-1h-6-chlorobenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,o-6-chloro-1-hydrocibenzotriazol-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate,2-6-chloro-1h-benzo d 1,2,3 triazol-1-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v,pubchem12740,acmc-20aj2u
42624899
[(6-chlorobenzotriazol-1-yl)oxy-(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
CN(C)C(=[N+](C)C)ON1C2=C(C=CC(=C2)Cl)N=N1.F[P-](F)(F)(F)(F)F
Melting Point 185.0°C to 190.0°C
Infrared Spectrum Authentic
Quantity 10g
Loss on Drying 0.5% max.
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 413.69
Percent Purity 98%
Chemical Name or Material O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning