Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog Number p-7028678
Click to view available options
Quantity:
1 g
5 g
25 g
148893-10-1
C10H15F6N6OP
380.24
MFCD00274639
JNWBBCNCSMBKNE-UHFFFAOYSA-N
hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide
9886157
[dimethylamino(triazolo[4,5-b]pyridin-3-yloxy)methylidene]-dimethylazanium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C
148893-10-1
C10H15F6N6OP
380.24
MFCD00274639
JNWBBCNCSMBKNE-UHFFFAOYSA-N
hatu, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-7-azabenzotriazol-1-yl-n,n,n,n-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethyluronium hexafluorophosphate v, dimethylamino triazolo 4,5-b pyridin-3-yloxy methylene-dimethyl-ammonium hexafluorophosphate, 2-7-aza-1h-benzotriazole-1-yl-1,1,3,3-tetramethyl uronium hexafluorophosphate, 2-3h-1,2,3 triazolo 4,5-b pyridin-3-yl-1,1,3,3-tetramethylisouronium hexafluorophosphate v, n-dimethylamino 3h-1,2,3 triazolo 4,5-b pyridin-3-yloxy methylene-n-methylmethanaminium hexafluorophosphate, n-dimethylamino-1h-1,2,3-triazolo-4,5-b pyridin-1-ylmethylene-n-methylmethanaminium hexafluorophosphate n-oxide
9886157
[dimethylamino(triazolo[4,5-b]pyridin-3-yloxy)methylidene]-dimethylazanium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C

O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.

Solubility
Soluble in acetonitrile. Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and highly acidic or alkaline materials.
TRUSTED_SUSTAINABILITY
Melting Point ∼185°C (decomposition)
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile. Insoluble in water.
Formula Weight 380.23
Percent Purity 99%
Chemical Name or Material O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
RTECSNumber XZ5633000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit