Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Acetanisidide 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
93-26-5
C9H11NO2
165.192
MFCD00026117
FGOFNVXHDGQVBG-UHFFFAOYSA-N
7135
N-(2-methoxyphenyl)acetamide
CC(=O)NC1=CC=CC=C1OC
93-26-5
C9H11NO2
165.192
MFCD00026117
FGOFNVXHDGQVBG-UHFFFAOYSA-N
7135
N-(2-methoxyphenyl)acetamide
CC(=O)NC1=CC=CC=C1OC
Color White
Quantity 500g
Melting Point 87°C
Boiling Point 305°C
Formula Weight 165.19
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material o-Acetanisidide

EINECSNumber : (3)-0690

RTECSNumber : AE8280000

TSCA : Yes